Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyridin-3-yl)-4,5-dihydrooxazole 97%

Supplier: AMBEED, INC
Warning

A220351-100MG A220351-1G A220351-250MG A220351-5G
77165-188EA 39 USD InStock
77165-188 77165-190 77165-192 77165-194
2-(Pyridin-3-yl)-4,5-dihydrooxazole 97%
2-(Pyridin-3-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD00183228
CAS Number: 40055-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty