Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3,5-dimethoxyaniline 98%

Supplier: AMBEED, INC
Warning

A221478-100MG A221478-1G A221478-250MG A221478-25G A221478-5G
77165-224EA 7 USD InStock
77165-224 77165-226 77165-228 77165-230 77165-232
2,6-Dichloro-3,5-dimethoxyaniline 98%
2,6-Dichloro-3,5-dimethoxyaniline
Precaución: for Research Use Only


Formula: C₈H₉Cl₂NO₂
MW: 222.06 g/mol
MDL Number: MFCD21648506
CAS Number: 872509-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty