Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorophenyl acetate 98%

Supplier: AMBEED, INC
Danger

A222532-25G A222532-5G
77192-934EA 18 USD InStock
77192-934 77192-936
4-Fluorophenyl acetate 98%
4-Fluorophenyl acetate
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00041361
CAS Number: 405-51-6
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty