Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)butanoic acid 98%

Supplier: AMBEED, INC
Warning

A223181-1G A223181-250MG A223181-5G
77165-270EA 92 USD InStock
77165-270 77165-272 77165-274
4-(Dimethylamino)butanoic acid 98%
4-(Dimethylamino)butanoic acid
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂
MDL Number: MFCD00044628
CAS Number: 693-11-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty