Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Nd-trityl-L-β-homoglutamine 98%

Supplier: AMBEED, INC
Warning

A226858-100MG A226858-1G A226858-250MG
77193-018EA 43 USD InStock
77193-018 77193-020 77193-022
Fmoc-Nd-trityl-L-β-homoglutamine 98%
Fmoc-Nd-trityl-L-beta-homoglutamine
Precaución: for Research Use Only


Formula: C₄₀H₃₆N₂O₅
MDL Number: MFCD03427580
CAS Number: 401915-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty