Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-3-oxopropanoate 98%

Supplier: AMBEED, INC
Warning

A228215-100G A228215-10G A228215-1G A228215-25G A228215-5G
77165-464EA 384 USD InStock
77165-464 77165-466 77165-468 77165-470 77165-472
Ethyl-3-amino-3-oxopropanoate 98%
Ethyl-3-amino-3-oxopropanoate
Precaución: for Research Use Only


Formula: C₅H₉NO₃
MDL Number: MFCD00043150
CAS Number: 7597-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty