Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A230764-1G A230764-250MG A230764-5G
77193-120EA 28.84 USD InStock
77193-120 77193-122 77193-124 77193-126
1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 98%
1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₁ClF₃NO
MDL Number: MFCD23379654
CAS Number: 1373925-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty