Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-nitro-1H-pyrrole-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A233356-100MG A233356-1G A233356-250MG
77165-598EA 18 USD InStock
77165-598 77165-600 77165-602
Methyl 5-nitro-1H-pyrrole-2-carboxylate 95%
Methyl 5-nitro-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₆N₂O₄
MDL Number: MFCD09743413
CAS Number: 13138-73-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty