Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(2-Bromophenyl)ethanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A234196-100MG A234196-1G A234196-250MG A234196-5G
77165-626EA 5 USD InStock
77165-626 77165-628 77165-630 77165-632
(R)-1-(2-Bromophenyl)ethanamine hydrochloride 97%
(R)-1-(2-Bromophenyl)ethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁BrClN
MW: 236.54 g/mol
MDL Number: MFCD09997782
CAS Number: 1187931-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty