Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-1-(2-nitrovinyl)benzene 95%

Supplier: AMBEED, INC
Danger

A235746-10G A235746-250MG A235746-5G
77193-192EA 189 USD InStock
77193-192 77193-194 77193-196
2,4-Dichloro-1-(2-nitrovinyl)benzene 95%
2,4-Dichloro-1-(2-nitrovinyl)benzene
Precaución: for Research Use Only


Formula: C₈H₅Cl₂NO₂
MDL Number: MFCD00053063
CAS Number: 18984-21-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty