Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Dimethoxymethylpyrimidine 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A239362-10G A239362-1G A239362-5G A239362-100G A239362-25G
77165-728EA 44 USD InStock
77165-728 77165-730 77165-732 77683-723 77683-724
4-Dimethoxymethylpyrimidine 98%, Ambeed.Inc
4-Dimethoxymethylpyrimidine
Precaución: For research use only.


Formula: C₇H₁₀N₂O₂
MDL Number: MFCD06738842
CAS Number: 25746-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty