Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-(2-bromo-4-nitrophenyl)-2-methylmalonate 98%

Supplier: AMBEED, INC
Warning

A241514-1G A241514-250MG
77165-770EA 17.51 USD InStock
77165-770 77165-772 77165-774
Diethyl 2-(2-bromo-4-nitrophenyl)-2-methylmalonate 98%
Diethyl 2-(2-bromo-4-nitrophenyl)-2-methylmalonate
Precaución: for Research Use Only


Formula: C₁₄H₁₆BrNO₆
MDL Number: MFCD28902467
CAS Number: 945244-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty