Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydroxybenzhydrazide 95%

Supplier: AMBEED, INC
Warning

A241739-1G A241739-25G A241739-5G
77193-348EA 17 USD InStock
77193-348 77193-350 77193-352
3,4-Dihydroxybenzhydrazide 95%
3,4-Dihydroxybenzhydrazide
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MW: 168.15
MDL Number: MFCD00017064
CAS Number: 39635-11-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty