Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Indandione 95%

Supplier: AMBEED, INC
Warning

A241992-100G A241992-10G A241992-1G A241992-25G A241992-5G
77193-354EA 135 USD InStock
77193-354 77193-356 77193-358 77193-360 77193-362
1,3-Indandione 95%
1,3-Indandione
Precaución: for Research Use Only


Formula: C₉H₆O₂
MDL Number: MFCD00003779
CAS Number: 606-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty