Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-Thr(tBu)-OH 97%

Supplier: AMBEED, INC
Warning

A242236-100G A242236-10G A242236-1G A242236-25G A242236-5G
77193-372EA 390 USD InStock
77193-372 77193-374 77193-376 77193-378 77193-380 77193-382
Z-Thr(tBu)-OH 97%
Z-Thr(tBu)-OH
Precaución: for Research Use Only


Formula: C₁₆H₂₃NO₅
MDL Number: MFCD06248909
CAS Number: 16966-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty