Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,6-Tetrafluoro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A242527-1G A242527-25G A242527-5G
77193-388EA 10 USD InStock
77193-388 77193-390 77193-392
α,α,α,6-Tetrafluoro-o-toluic acid 98%
α,α,α,6-Tetrafluoro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₄F₄O₂
MW: 208.11
MDL Number: MFCD00040981
CAS Number: 32890-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty