Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-β-Ionone 95%

Supplier: AMBEED, INC
Warning

A242574-100G A242574-25G A242574-500G
77165-800EA 21 USD InStock
77165-800 77165-802 77165-804
(E)-β-Ionone 95%
(E)-β-Ionone
Precaución: for Research Use Only


Formula: C₁₃H₂₀O
MDL Number: MFCD00001549
CAS Number: 79-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty