Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptane-1,7-diamine, N-BOC protected 97%

Supplier: AMBEED, INC
Warning

A243091-100MG A243091-1G A243091-250MG A243091-5G A243091-25G
77193-408EA 40 USD InStock
77193-408 77193-410 77193-412 77193-414 77746-370
Heptane-1,7-diamine, N-BOC protected 97%
Heptane-1,7-diamine, N-BOC protected
Precaución: for Research Use Only


Formula: C₁₂H₂₆N₂O₂
MDL Number: MFCD02094498
CAS Number: 99733-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty