Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A244121-10G A244121-1G A244121-250MG A244121-5G
77165-826EA 100 USD InStock
77165-826 77165-828 77165-830 77165-832
5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride 95%
5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₇ClN₄
MDL Number: MFCD26407488
CAS Number: 23350-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty