Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diiodopropane 97% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with copper chip

A245289-100G A245289-10G A245289-25G A245289-500G A245289-5G
77193-480EA 67 USD InStock
77193-480 77193-482 77193-484 77193-486 77193-488
1,3-Diiodopropane 97% stabilised
1,3-Diiodopropane
Precaución: for Research Use Only


Formula: C₃H₆I₂
MW: 295.89 g/mol
Boiling Pt: 228
Melting Pt: -20
MDL Number: MFCD00001097
CAS Number: 627-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty