Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(chloromethyl)tetramethyldisiloxane 95%

Supplier: AMBEED, INC
Warning

A245510-100G A245510-25G A245510-500G A245510-5G
77193-496EA 89 USD InStock
77193-496 77193-498 77193-500 77193-502
1,3-Bis(chloromethyl)tetramethyldisiloxane 95%
1,3-Bis(chloromethyl)tetramethyldisiloxane
Precaución: for Research Use Only


Formula: C₆H₁₆Cl₂OSi₂
MDL Number: MFCD00000877
CAS Number: 2362-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty