Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BAPTA tetrapotassium salt 97%

Supplier: AMBEED, INC
Warning

A247327-100MG A247327-10G A247327-1G A247327-250MG A247327-5G
77193-536EA 23 USD InStock
77193-536 77193-538 77193-540 77193-542 77193-544
BAPTA tetrapotassium salt 97%
BAPTA tetrapotassium salt
Precaución: for Research Use Only


Formula: C₂₂H₂₀N₂O₁₀K₄
MDL Number: MFCD00070080
CAS Number: 73630-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty