Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-2-chloropyridine 97%

Supplier: AMBEED, INC
Warning

A247765-100G A247765-1G A247765-25G A247765-5G
77193-564EA 420 USD InStock
77193-564 77193-566 77193-568 77193-570
4-Acetyl-2-chloropyridine 97%
4-Acetyl-2-chloropyridine
Precaución: for Research Use Only


Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD07699406
CAS Number: 23794-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty