Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-2-bromo-2-methylmalonate 98%

Supplier: AMBEED, INC
Danger

A248135-10G A248135-1G A248135-25G A248135-5G
77193-586EA 13 USD InStock
77193-586 77193-588 77193-590 77193-592
Diethyl-2-bromo-2-methylmalonate 98%
Diethyl-2-bromo-2-methylmalonate
Precaución: for Research Use Only


Formula: C₈H₁₃BrO₄
MW: 253.09 g/mol
MDL Number: MFCD00009125
CAS Number: 29263-94-3
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty