Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propyl benzenesulfonate 97%

Supplier: AMBEED, INC
Warning

A248723-100MG A248723-10G A248723-1G A248723-250MG A248723-25G A248723-5G
77193-612EA 5 USD InStock
77193-612 77193-614 77193-616 77193-618 77193-620 77193-622
Propyl benzenesulfonate 97%
Propyl benzenesulfonate
Precaución: for Research Use Only


Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
MDL Number: MFCD00996025
CAS Number: 80-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty