Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethoxy)phenylacetylene 98%

Supplier: AMBEED, INC
Danger

A250840-100MG A250840-250MG
77165-958EA 23 USD InStock
77165-958 77165-960
3-(Trifluoromethoxy)phenylacetylene 98%
3-(Trifluoromethoxy)phenylacetylene
Precaución: for Research Use Only


Formula: C₉H₅F₃O
MW: 186.131 g/mol
MDL Number: MFCD08458148
CAS Number: 866683-57-0
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty