Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoro-2-methylpropiophenone 95%

Supplier: AMBEED, INC
Warning

A254410-100MG A254410-1G A254410-250MG A254410-5G
77193-710EA 11 USD InStock
77193-710 77193-712 77193-714 77193-716
4'-Fluoro-2-methylpropiophenone 95%
4'-Fluoro-2-methylpropiophenone
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO
MDL Number: MFCD04038305
CAS Number: 26393-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty