Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-3-Aminocyclopentanecarboxylic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A257843-100MG A257843-250MG
77166-140EA 50 USD InStock
77166-140 77166-142
trans-3-Aminocyclopentanecarboxylic acid hydrochloride 97%
trans-3-Aminocyclopentanecarboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClNO₂
MDL Number: MFCD23106174
CAS Number: 1392803-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty