Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azepane-2,4-dione 98%

Supplier: AMBEED, INC
Warning

A264150-100MG A264150-10G A264150-1G A264150-250MG A264150-5G
77166-270EA 51 USD InStock
77166-270 77166-272 77166-274 77166-276 77166-278
Azepane-2,4-dione 98%
Azepane-2,4-dione
Precaución: for Research Use Only


Formula: C₆H₉NO₂
MDL Number: MFCD19217055
CAS Number: 29520-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty