Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%

Supplier: AMBEED, INC
Danger

A267610-100MG A267610-1G A267610-250MG A267610-5G
77193-976EA 11 USD InStock
77193-976 77193-978 77193-980 77193-982
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride
Precaución: for Research Use Only


Formula: C₆H₉ClN₂O₂S
MW: 208.6700 g/mol
MDL Number: MFCD02180944
CAS Number: 59340-27-1
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty