Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5-Triiodobenzaldehyde 97%

Supplier: AMBEED, INC
Danger

A269801-100MG A269801-1G A269801-250MG
77166-392EA 34 USD InStock
77166-392 77166-394 77166-396
2,3,5-Triiodobenzaldehyde 97%
2,3,5-Triiodobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃I₃O
MDL Number: MFCD08703354
CAS Number: 477534-99-9
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty