Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Methoxyphenyl)-1H-pyrrole-2,5-dione 95%

Supplier: AMBEED, INC
Danger

A269942-100MG A269942-1G A269942-250MG A269942-5G
77193-988EA 14 USD InStock
77193-988 77193-990 77193-992 77193-994
1-(4-Methoxyphenyl)-1H-pyrrole-2,5-dione 95%
1-(4-Methoxyphenyl)-1H-pyrrole-2,5-dione
Precaución: for Research Use Only


Formula: C₁₁H₉NO₃
MDL Number: MFCD00022579
CAS Number: 1081-17-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty