Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-fluoropicolinonitrile 95%

Supplier: AMBEED, INC
Warning

A273736-100MG A273736-250MG A273736-25G A273736-5G
77166-512EA 26 USD InStock
77166-512 77166-514 77166-516 77166-518
6-Chloro-3-fluoropicolinonitrile 95%
6-Chloro-3-fluoropicolinonitrile
Precaución: for Research Use Only


Formula: C₆H₂ClFN₂
MDL Number: MFCD18250470
CAS Number: 1207609-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty