Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Methoxyphenyl)imidazole 98%

Supplier: AMBEED, INC
Warning

A275346-100MG A275346-1G A275346-250MG
77194-052EA 12 USD InStock
77194-052 77194-054 77746-489
1-(4-Methoxyphenyl)imidazole 98%
1-(4-Methoxyphenyl)imidazole
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00060492
CAS Number: 10040-95-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty