Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline-6-amine 98%

Supplier: AMBEED, INC
Warning

A278031-100MG A278031-10G A278031-1G A278031-250MG A278031-5G
77166-590EA 7 USD InStock
77166-590 77166-592 77166-594 77166-596 77166-598
Isoquinoline-6-amine 98%
Isoquinoline-6-amine
Precaución: for Research Use Only


Formula: C₉H₈N₂
MDL Number: MFCD04114862
CAS Number: 23687-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty