Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4''-(Pentyloxy)-[1,1':4',1''-terphenyl]-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A279274-10G A279274-1G A279274-250MG A279274-25G A279274-5G
77166-618EA 257.01 USD InStock
77166-618 77166-620 77166-622 77166-624 77166-626
4''-(Pentyloxy)-[1,1':4',1''-terphenyl]-4-carboxylic acid 97%
4''-(Pentyloxy)-[1,1':4',1''-terphenyl]-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₄H₂₄O₃
MDL Number: MFCD17019206
CAS Number: 158938-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty