Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitroisatoic anhydride 95%

Supplier: AMBEED, INC
Warning

A281431-10G A281431-1G A281431-250MG A281431-5G
77194-148EA 27 USD InStock
77194-148 77194-150 77194-152 77194-154
4-Nitroisatoic anhydride 95%
4-Nitroisatoic anhydride
Precaución: for Research Use Only


Formula: C₈H₄N₂O₅
MDL Number: MFCD00798220
CAS Number: 63480-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty