Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophthalic anhydride 97%

Supplier: AMBEED, INC
Warning

A281493-100G A281493-25G A281493-500G
77194-156EA 33 USD InStock
77194-156 77194-158 77194-160
4-Bromophthalic anhydride 97%
4-Bromophthalic anhydride
Precaución: for Research Use Only


Formula: C₈H₃BrO₃
MW: 227.01 g/mol
MDL Number: MFCD00191323
CAS Number: 86-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty