Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(3-amino-4-fluorophenyl)acetate 98%

Supplier: AMBEED, INC
Warning

A285890-100MG A285890-250MG A285890-5G
77166-688EA 10 USD InStock
77166-688 77166-690 77166-692
Ethyl 2-(3-amino-4-fluorophenyl)acetate 98%
Ethyl 2-(3-amino-4-fluorophenyl)acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂FNO₂
MW: 197.20 g/mol
MDL Number: MFCD16620934
CAS Number: 858972-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty