Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A286346-1G A286346-250MG A286346-5G
77166-702EA 99 USD InStock
77166-702 77166-704 77166-706
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MDL Number: MFCD02251327
CAS Number: 30384-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty