Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A288370-1G A288370-250MG A288370-5G
77166-750EA 97 USD InStock
77166-750 77166-752 77166-754
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride 95%
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClFNO₂
MDL Number: MFCD19105591
CAS Number: 1001354-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty