Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-3-ethylpyrazine 98%

Supplier: AMBEED, INC
Warning

A291800-10G A291800-5G
77194-336EA 41 USD InStock
77194-336 77194-338
2-Acetyl-3-ethylpyrazine 98%
2-Acetyl-3-ethylpyrazine
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O
MDL Number: MFCD00038028
CAS Number: 32974-92-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty