Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-(tert-Butoxycarbonyl)-1H-indol-4-yl)boronic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A293921-1G A293921-250MG A293921-5G A293921-100MG
77166-840EA 210 USD InStock
77166-840 77166-842 77166-844 77683-880
(1-(tert-Butoxycarbonyl)-1H-indol-4-yl)boronic acid 95%, Ambeed.Inc
(1-(tert-Butoxycarbonyl)-1H-indol-4-yl)boronic acid
Precaución: For research use only.


Formula: C₁₃H₁₆BNO₄
MW: 261.08 g/mol
MDL Number: MFCD28130132
CAS Number: 2102451-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty