Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzo[d]oxazol-2-yl)acetonitrile 96%

Supplier: AMBEED, INC
Warning

A294472-100MG A294472-250MG A294472-5G
77194-394EA 17 USD InStock
77194-394 77194-396 77194-398
2-(Benzo[d]oxazol-2-yl)acetonitrile 96%
2-(Benzo[d]oxazol-2-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₉H₆N₂O
MDL Number: MFCD00160031
CAS Number: 15344-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty