Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrobenzenesulfonic acid hydrate 97%

Supplier: AMBEED, INC
Danger

A295509-1G A295509-250MG A295509-5G
77166-896EA 31 USD InStock
77166-896 77166-898 77166-900
3-Nitrobenzenesulfonic acid hydrate 97%
3-Nitrobenzenesulfonic acid hydrate
Precaución: for Research Use Only


Formula: C₆H₇NO₆S
MDL Number: MFCD00064383
CAS Number: 98-47-5
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty