Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl amino(2-chlorophenyl)acetate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A296471-1G A296471-250MG A296471-5G
77194-400EA 23 USD InStock
77194-400 77194-402 77194-404
Methyl amino(2-chlorophenyl)acetate hydrochloride 97%
Methyl amino(2-chlorophenyl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₀ClNO₂·ClH
MDL Number: MFCD16661100
CAS Number: 141109-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty