Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-fluoroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A298202-100MG A298202-250MG
77194-418EA 28 USD InStock
77194-418 77194-420
5-Bromo-6-fluoroisoquinoline 97%
5-Bromo-6-fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₅BrFN
MDL Number: MFCD20527917
CAS Number: 1239463-43-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty