Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SKLB1002 ≥98%

Supplier: AMBEED, INC
Warning

A300860-100MG A300860-10MG A300860-1MG A300860-25MG A300860-5MG
77194-450EA 77 USD InStock
77194-450 77194-452 77194-454 77194-456 77194-458
SKLB1002 ≥98%
SKLB1002
Precaución: for Research Use Only


Formula: C₁₃H₁₂N₄O₂S₂
MDL Number: MFCD27938707
CAS Number: 1225451-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty