Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-5-methylthiophene 96%

Supplier: AMBEED, INC
Warning

A303223-100G A303223-1G A303223-25G A303223-5G
77194-480EA 82 USD InStock
77194-480 77194-482 77194-484 77194-486
2-Acetyl-5-methylthiophene 96%
2-Acetyl-5-methylthiophene
Precaución: for Research Use Only


Formula: C₇H₈OS
MDL Number: MFCD00014529
CAS Number: 13679-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty