Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ISX9 98%

Supplier: AMBEED, INC
Warning

A305535-10MG A305535-1MG A305535-50MG A305535-5MG
77194-510EA 23 USD InStock
77194-510 77194-512 77194-514 77194-516
ISX9 98%
ISX9
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₂S
MDL Number: MFCD04133937
CAS Number: 832115-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty